Skip to Content
Merck
All Photos(1)

Key Documents

760862

Sigma-Aldrich

Poly(ethylene glycol), 8 hydroxyl dendron, generation 2

average Mn 6,700

Synonym(s):

Bis-MPA poly(ethylene glycol), Bis[2,2-bis(hydroxymethyl)propanoic acid] PEG, Dendron functionalized PEG, generation 2, PFD-G2-PEG6k-OH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

PEG average Mn 6,000 (n~130; esterified with 8 hydroxyl groups)
average Mn 6,700

feature

generation 2

mp

48-53 °C

functional group

hydroxyl

Looking for similar products? Visit Product Comparison Guide

General description

Dendrimers are symmetrically branched polymer structures, monodispersed macromolecules. They possess uniform size, water solubiltity, and modifiable surface functionality. Cross linking it with polyethylene glycol results in very high water content.

Application

1. Dendritic nanodevices for therapeutic applications.
2. Surface modified for targeting, imaging and drug delivery
3. Antibody-dendrimer conjugate
4. Anti viral drugs
5. Tissue repair scaffolds

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anil K Patri et al.
Current opinion in chemical biology, 6(4), 466-471 (2002-07-23)
Dendrimers are synthetic, highly branched, mono-disperse macromolecules of nanometer dimensions. Started in the mid-1980s, the research investigations into the synthetic methodology, physical and chemical properties of these macromolecules are increasing exponentially with growing interest in this field. Potential applications for
Degradable Poly(ethylene glycol) Hydrogels
for 2D and 3D Cell Culture
Kasko A
Material Matters, 6(3), 67-75 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service