Skip to Content
Merck
All Photos(1)

Key Documents

75653

Sigma-Aldrich

(1R,5S)-3-Oxabicyclo[3.3.0]oct-6-en-2-one

≥95.0% (GC)

Synonym(s):

(3aS,6aR)-3,3a,6,6a-Tetrahydro-1H-cyclopenta[c]furan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
Beilstein:
4661660
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95.0% (GC)

form

liquid

optical activity

[α]20/D 53±2°, c = 0.5% in methanol

functional group

ester

storage temp.

2-8°C

SMILES string

[H][C@@]12COC(=O)[C@]1([H])CC=C2

InChI

1S/C7H8O2/c8-7-6-3-1-2-5(6)4-9-7/h1-2,5-6H,3-4H2/t5-,6-/m1/s1

InChI key

XYRFGFRTVXZAIU-PHDIDXHHSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chiral building block; e.g. for pheromone synthesis[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Lebreton et al.
Tetrahedron, 53, 145-145 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service