Skip to Content
Merck
All Photos(1)

Documents

755427

Sigma-Aldrich

4,6-Dichloro-2-(methylthio)pyrimidine-5-carbonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl3N2OS
CAS Number:
Molecular Weight:
257.52
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

72-77 °C

SMILES string

CSc1nc(Cl)c(C(Cl)=O)c(Cl)n1

InChI

1S/C6H3Cl3N2OS/c1-13-6-10-3(7)2(5(9)12)4(8)11-6/h1H3

InChI key

MHZGWIOWCZZQNF-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service