Skip to Content
Merck
All Photos(1)

Documents

75401

Sigma-Aldrich

HypoGel® 200 CHO

extent of labeling: ~0.8 mmol/g loading

Synonym(s):

O-[2-(6-Oxocaproylamino)ethyl]tetra(oxyethylene) polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

crystals

extent of labeling

~0.8 mmol/g loading

particle size

110-150 μm

application(s)

peptide synthesis

storage temp.

2-8°C

Legal Information

HypoGel is a registered trademark of Rapp Polymere GmbH

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service