Skip to Content
Merck
All Photos(1)

Documents

751340

Sigma-Aldrich

4-Stilbenecarboxylic acid

97%

Synonym(s):

4-(2-Phenylethenyl)benzoic acid, 4-Styrylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O2
CAS Number:
Molecular Weight:
224.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

248-253 °C

SMILES string

OC(=O)c1ccc(\C=C\c2ccccc2)cc1

InChI

1S/C15H12O2/c16-15(17)14-10-8-13(9-11-14)7-6-12-4-2-1-3-5-12/h1-11H,(H,16,17)/b7-6+

InChI key

IAGXTPCOGVFRSQ-VOTSOKGWSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakub Treml et al.
Biomolecules, 9(9) (2019-09-12)
The stilbenoids, a group of naturally occurring phenolic compounds, are found in a variety of plants, including some berries that are used as food or for medicinal purposes. They are known to be beneficial for human health as anti-inflammatory, chemopreventive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service