Skip to Content
Merck
All Photos(1)

Key Documents

746975

Sigma-Aldrich

Propylene oxide-1,3-13C2

99 atom % 13C, 98% (CP), contains hydroquinone as a stabilizer

Synonym(s):

13C Labeled Propylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2CH6O
Molecular Weight:
60.06
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

liquid

quality

contains hydroquinone as a stabilizer

mass shift

M+2

SMILES string

[13CH3]C1[13CH2]O1

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3/i1+1,2+1

InChI key

GOOHAUXETOMSMM-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-162.4 °F - closed cup

Flash Point(C)

-108 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service