Skip to Content
Merck
All Photos(2)

Key Documents

746908

Sigma-Aldrich

Methyl 2,3,4-triacetate-α-D-glucopyranoside

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O9
CAS Number:
Molecular Weight:
320.29
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

optical activity

[α]22/D +148°, c = 1% in DMSO

mp

108-113 °C

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](OC)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C13H20O9/c1-6(15)19-10-9(5-14)22-13(18-4)12(21-8(3)17)11(10)20-7(2)16/h9-14H,5H2,1-4H3/t9-,10-,11+,12-,13+/m1/s1

InChI key

OBVFJYUDIWPVKD-LBELIVKGSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mechanistic study of the reaction of phenyl isocyanate with methyl 2,3,4-tri-O-acetyl-alpha-D-glucopyranoside.
S Knapp et al.
Carbohydrate research, 203(2), 290-295 (1990-08-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service