Skip to Content
Merck
All Photos(1)

Key Documents

746894

Sigma-Aldrich

2-Acetamido-3-acetyl-4.6-di-O-benzylidene-2-deoxy-α-D-galactopyranose

95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO7
Molecular Weight:
351.35
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

95% (HPLC)

form

solid

mp

192-197 °C

storage temp.

−20°C

SMILES string

O[C@@H]1[C@H](NC(C)=O)[C@@H](OC(C)=O)[C@@H]2[C@@H](COC(C3=CC=CC=C3)O2)O1

InChI

1S/C17H21NO7/c1-9(19)18-13-15(23-10(2)20)14-12(24-16(13)21)8-22-17(25-14)11-6-4-3-5-7-11/h3-7,12-17,21H,8H2,1-2H3,(H,18,19)/t12-,13-,14+,15-,16+,17?/m1/s1

InChI key

OPEPHRZBCPGJGP-QNGFNYMWSA-N

Application

Intermediate used in the preparation of TN-Ser/Thr

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service