Skip to Content
Merck
All Photos(1)

Key Documents

745316

Sigma-Aldrich

4-(Azidomethyl)benzonitrile solution

~0.5 M in tert-butyl methyl ether, ≥97.0% (HPLC)

Synonym(s):

α-Azido-4-cyanotoluene solution, 4-Cyanobenzyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N4
CAS Number:
Molecular Weight:
158.16
Beilstein:
4978502
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

liquid

reaction suitability

reaction type: click chemistry

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

2-8°C

SMILES string

[N-]=[N+]=NCc1ccc(cc1)C#N

InChI

1S/C8H6N4/c9-5-7-1-3-8(4-2-7)6-11-12-10/h1-4H,6H2

InChI key

GWVIVHINAVAKKV-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F

Flash Point(C)

-30 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted benzyne-click chemistry: preparation of 1H-benzo[d][1,2,3]triazoles
Ankatia, H. and Biehl, E.
Tetrahedron Letters, 50, 4677-4682 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service