Skip to Content
Merck
All Photos(1)

Documents

745219

Sigma-Aldrich

(5H-Dibenzo[a,d]cyclohepten-5-yl)diphenylphosphine

≥96.5% (elemental analysis)

Synonym(s):

(5H-Dibenzo[a,d]cyclohepten-5-yl)diphenylphosphane, 5-(Diphenylphosphanyl)dibenzo[a,d]cycloheptene, troppPh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21P
CAS Number:
Molecular Weight:
376.43
Beilstein:
12068554
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (elemental analysis)

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

loss

≤1.0% loss on drying

functional group

phosphine

SMILES string

P(C1=CC=CC=C1)(C2=CC=CC=C2)C3C(C=CC=C4)=C4C=CC5=C3C=CC=C5

InChI

1S/C27H21P/c1-3-13-23(14-4-1)28(24-15-5-2-6-16-24)27-25-17-9-7-11-21(25)19-20-22-12-8-10-18-26(22)27/h1-20,27H

InChI key

KZCMNDBHBVBRLY-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electromeric rhodium radical complexes.
Florian Frank Puschmann et al.
Angewandte Chemie (International ed. in English), 49(2), 385-389 (2009-12-04)
Torsten Büttner et al.
Chemical communications (Cambridge, England), 24(24), 2820-2821 (2004-12-16)
A combination of the rigid bis(5H-dibenzo[a,d]cyclohepten-5-yl)amine (trop(2)NH) and 5-amino-5H-dibenzo[a,d]cycloheptene (tropNH(2)) ligand allowed the synthesis of the stable pentacoordinated 18 electron amine olefin rhodium(i) complex [Rh(trop(2)NH)(tropNH(2))]O(3)SCF(3)(); this complex can be cleanly deprotonated [pK(a)(DMSO) = 20.6(1)] to the corresponding amide [Rh(trop(2)N)(tropNH(2))](6) which
Chiral Rhodium(I) and Iridium(I) Amino-Olefin Complexes:? pKa, N-H Bond Dissociation Energy, and Catalytic Transfer Hydrogenation
Maire, P., et al.
Organometallics, 24, 3207-3218 (2005)
Ethanol as hydrogen donor: highly efficient transfer hydrogenations with rhodium(I) amides.
Theo Zweifel et al.
Angewandte Chemie (International ed. in English), 47(17), 3245-3249 (2008-03-19)
Heterolytic splitting of hydrogen with rhodium(I) amides.
Pascal Maire et al.
Angewandte Chemie (International ed. in English), 44(39), 6318-6323 (2005-09-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service