Skip to Content
Merck
All Photos(1)

Key Documents

736708

Sigma-Aldrich

Ethyl-d5 methyl-d3 carbonate

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2OCOOCD3
CAS Number:
Molecular Weight:
112.15
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

liquid

mass shift

M+8

SMILES string

[2H]C([2H])([2H])OC(=O)OC([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C4H8O3/c1-3-7-4(5)6-2/h3H2,1-2H3/i1D3,2D3,3D2

InChI key

JBTWLSYIZRCDFO-AUOAYUKBSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable Liquids

WGK

WGK 1

Flash Point(F)

80.1 °F

Flash Point(C)

26.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service