Skip to Content
Merck
All Photos(1)

Documents

736538

Sigma-Aldrich

Diethyl vinyl orthoformate

97%

Synonym(s):

(Diethoxymethoxy)ethene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.401

density

0.822 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCOC(OCC)OC=C

InChI

1S/C7H14O3/c1-4-8-7(9-5-2)10-6-3/h4,7H,1,5-6H2,2-3H3

InChI key

AOYJISNQMSDAHR-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

64.0 °F

Flash Point(C)

17.8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service