Skip to Content
Merck
All Photos(2)

Key Documents

736317

Sigma-Aldrich

Lithium tetrachlorogallate

anhydrous, beads, −10 mesh, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiGaCl4
CAS Number:
Molecular Weight:
218.48
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99% trace metals basis

form

beads

particle size

−10 mesh

mp

175 °C (lit.)

storage temp.

2-8°C

SMILES string

[Li+].Cl[Ga-](Cl)(Cl)Cl

InChI

1S/4ClH.Ga.Li/h4*1H;;/q;;;;+3;+1/p-4

InChI key

KMBVSAMILLNTNQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Russell, P. G.; Reddy, T. B.
Proc. 37th Power Sources Conf., 365-365 (1996)
West, W. C.; Shevade, A.; Soler, J.; Kulleck, J.; Smart, M.
Journal of the Electrochemical Society, 157, A571-A571 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service