Skip to Content
Merck
All Photos(1)

Key Documents

736112

Sigma-Aldrich

Capstone® FS-66

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

form

solid

mp

38-52 °C

General description

TM=["Capstone"] FS-66 (FS-66) is an anionic fluorosurfactant that is used to reduce the surface tension of solutions and it affects the uniformity and morphology of the polymeric matrix. It can also exhibit improved charge density and impact the electrostatic properties of the polymeric solutions.[1]

Application

TM=["Capstone"] FS-66 can be used as a surfactant in the preparation of polyvinyl fluoride (PVDF) solution for 3D electrospinning on inkjet patterned paper and for 3D printing of piezoelectric materials.[2][3][4][5][6]

Legal Information

Capstone is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of uniform PVDF fibers under ultrasound exposure in presence of anionic surfactant.
Tarasova E, et al.
Journal of Electrostatics, 76, 39-47 (2015)
Near field sequentially electrospun three-dimensional piezoelectric fibers arrays for self-powered sensors of human gesture recognition.
Fuh YK and Wang BS
Nano Energy, 30(7), 677-683 (2016)
Near-Field Electrospun Piezoelectric Fibers as Sound-Sensing Elements.
Lee T, et al.
Polymer, 10(7), 692-692 (2018)
Self-powered active sensor with concentric topography of piezoelectric fibers.
Fuh YK, et al.
Nanoscale Research Letters, 12(1), 44-44 (2017)
A Review on 3D printing of piezoelectric materials
IOP Conference Series: Materials Science and Engineering, 455(1), 012046-012046 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service