Skip to Content
Merck
All Photos(1)

Key Documents

733067

Sigma-Aldrich

2-Methoxy-4-methylaniline

98%

Synonym(s):

4-Methyl-o-anisidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO
CAS Number:
Molecular Weight:
137.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

31-33 °C

density

1.043 g/mL at 25 °C

SMILES string

COc1cc(C)ccc1N

InChI

1S/C8H11NO/c1-6-3-4-7(9)8(5-6)10-2/h3-5H,9H2,1-2H3

InChI key

CJJLEUQMMMLOFI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

219.2 °F

Flash Point(C)

104 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service