Skip to Content
Merck
All Photos(1)

Key Documents

732990

Sigma-Aldrich

7-Bromo-4-hydroxycoumarin

Synonym(s):

7-Bromo-4-hydroxy-2H-1-benzopyran-2-one, 7-Bromo-4-hydroxy-2H-chromen-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 104.00

CHF 104.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
CHF 104.00

About This Item

Empirical Formula (Hill Notation):
C9H5BrO3
CAS Number:
Molecular Weight:
241.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 104.00


Check Cart for Availability

Request a Bulk Order

form

solid

mp

244-249 °C

functional group

bromo
ester
ketone

SMILES string

OC1=CC(=O)Oc2cc(Br)ccc12

InChI

1S/C9H5BrO3/c10-5-1-2-6-7(11)4-9(12)13-8(6)3-5/h1-4,11H

InChI key

UJWIUPBUKQLTTJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service