Skip to Content
Merck
All Photos(1)

Key Documents

732680

Sigma-Aldrich

Methyl 1-ethyl-3-methyl-1H-pyrazole-5-carboxylate

Synonym(s):

1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O2
CAS Number:
Molecular Weight:
168.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.485

SMILES string

CCn1nc(C)cc1C(=O)OC

InChI

1S/C8H12N2O2/c1-4-10-7(8(11)12-3)5-6(2)9-10/h5H,4H2,1-3H3

InChI key

QQCDTXYUOXYOTM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

189.5 °F

Flash Point(C)

87.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service