728500
1-(4-Aminobutyl)-4-(2-methoxyphenyl)piperazine
97%
Synonym(s):
4-[4-(2-Methoxyphenyl)-1-piperazinyl]butylamine
About This Item
Assay
≥97.0% (HPLC)
97%
form
liquid
storage temp.
2-8°C
SMILES string
COc1ccccc1N2CCN(CCCCN)CC2
InChI
1S/C15H25N3O/c1-19-15-7-3-2-6-14(15)18-12-10-17(11-13-18)9-5-4-8-16/h2-3,6-7H,4-5,8-13,16H2,1H3
InChI key
UFJPFLDFMPVGRW-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service