Skip to Content
Merck
All Photos(1)

Key Documents

726788

Sigma-Aldrich

(R)-(+)-1-Phenylethanol

ChiPros®, produced by BASF, 98%

Synonym(s):

(+)-Methyl phenyl carbinol, (R)-(+)-α-Methylbenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
Beilstein:
2039798
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
form:
liquid
Assay:
≥97.5% (GC)
98%

grade

produced by BASF

Assay

≥97.5% (GC)
98%

form

liquid

optical purity

enantiomeric excess: ≥97.5%

bp

88-89 °C/10 mmHg (lit.)

mp

9-11 °C (lit.)

density

1.012 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](O)c1ccccc1

InChI

1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3/t7-/m1/s1

InChI key

WAPNOHKVXSQRPX-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachiko Sugimoto et al.
Phytochemistry, 108, 189-195 (2014-12-03)
Three aromatic glycosides (1-3), two sulfur and nitrogen-containing compound glucosides (4, 5), and one flavonoid glycoside (6) were isolated from the leaves of Ixora undulata. Their structures were established by extensive 1D, 2D NMR, and HRESIMS experiments, and structure 4

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service