Skip to Content
Merck
All Photos(1)

Documents

717061

Sigma-Aldrich

4,4-Difluoro-L-prolinamide hydrochloride

97%

Synonym(s):

(S)-4,4-Difluoropyrolidine-2-carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9ClF2N2O
CAS Number:
Molecular Weight:
186.59
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

optical activity

[α]/D −23±5°, c = 0.5 in H2O

SMILES string

Cl.NC(=O)[C@@H]1CC(F)(F)CN1

InChI

1S/C5H8F2N2O.ClH/c6-5(7)1-3(4(8)10)9-2-5;/h3,9H,1-2H2,(H2,8,10);1H/t3-;/m0./s1

InChI key

RRQDQYAEZGKHOB-DFWYDOINSA-N

Application

4,4-Difluoro-L-prolinamide hydrochloride can be used as a building block in the synthesis of cyanopyrrolidine based DPP-IV inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structure-activity relationships of potent 3-or 4-substituted-2-cyanopyrrolidine dipeptidyl peptidase IV inhibitors
Fukushima H, et al.
Bioorganic & Medicinal Chemistry, 12(23), 6053-6061 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service