Skip to Content
Merck
All Photos(1)

Documents

703133

Sigma-Aldrich

Dimethyl-13C2, d6 sulfoxide

99 atom % 13C, 99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2D6OS
Molecular Weight:
86.16
MDL number:
UNSPSC Code:
12191600
PubChem Substance ID:

isotopic purity

99 atom % 13C
99 atom % D

form

liquid

bp

189 °C

density

1.212 g/mL at 25 °C

mass shift

M+8

SMILES string

[2H][13C]([2H])([2H])S(=O)[13C]([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1+1D3,2+1D3

InChI key

IAZDPXIOMUYVGZ-KLFIIISESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service