Skip to Content
Merck
All Photos(1)

Key Documents

699799

Sigma-Aldrich

Poly(thiophene-3-[2-(2-methoxyethoxy)ethoxy]-2,5-diyl), sulfonated solution

2% in 1,2-propanediol/isopropanol/water, 3:2:1, electronic grade

Synonym(s):

Plexcore® OC RG-1100, Sulfonated polythiophene ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H12O6S2)X(C9H11O3S)Y
CAS Number:
UNSPSC Code:
12352103

grade

electronic grade

form

liquid

concentration

2% in 1,2-propanediol/isopropanol/water, 3:2:1

resistivity

25-250 Ω-cm

work function

−5.1-−5.2 eV

color

dark blue

pH

2.2-2.8

surface tension

35-38 dyn/cm

viscosity

7-13 cP, Brookfield

density

~0.98 g/mL at 25 °C

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Organic conductive ink suitable for spin coating applications and as hole injection layer in organic light emitting diodes (OLED).

Legal Information

Product of Plextronics, Inc. U.S. Patent number 6,166,172.
Plexcore is a registered trademark of Plextronics, Inc.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

90.0 °F

Flash Point(C)

32.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service