Skip to Content
Merck
All Photos(1)

Key Documents

698601

Sigma-Aldrich

3-Fluoro-4-(tributylstannyl)pyridine

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30FNSn
CAS Number:
Molecular Weight:
386.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98%

form

liquid

density

1.773 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccncc1F

InChI

1S/C5H3FN.3C4H9.Sn/c6-5-2-1-3-7-4-5;3*1-3-4-2;/h1,3-4H;3*1,3-4H2,2H3;

InChI key

KKOFZNIMGWDVQS-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service