Skip to Content
Merck
All Photos(1)

Key Documents

693464

Sigma-Aldrich

(S)-2-[3-[(1R,2R)-2-(Dipropylamino)cyclohexyl]thioureido]-N-isopropyl-3,3-dimethylbutanamide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H44N4OS
CAS Number:
Molecular Weight:
412.68
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

95%

optical activity

[α]22/D -61°, c = 1 in chloroform

SMILES string

CCCN(CCC)[C@@H]1CCCC[C@H]1NC(=S)N[C@H](C(=O)NC(C)C)C(C)(C)C

InChI

1S/C22H44N4OS/c1-8-14-26(15-9-2)18-13-11-10-12-17(18)24-21(28)25-19(22(5,6)7)20(27)23-16(3)4/h16-19H,8-15H2,1-7H3,(H,23,27)(H2,24,25,28)/t17-,18-,19-/m1/s1

InChI key

XULQYZJFCNMXTM-GUDVDZBRSA-N

Application

Ketone silylcyanation catalyst

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas E Fuerst et al.
Journal of the American Chemical Society, 127(25), 8964-8965 (2005-06-23)
The new chiral amino thiourea catalyst 3d promotes the highly enantioselective cyanosilylation of a wide variety of ketones. The hindered tertiary amine substituent plays a crucial role with regard to both stereoinduction and reactivity, suggesting a cooperative mechanism involving electrophile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service