Skip to Content
Merck
All Photos(2)

Key Documents

691992

Sigma-Aldrich

2-Nitrophenethyl bromide

97%

Synonym(s):

1-(2-Bromoethyl)-2-nitrobenzene, 2-(2-Bromoethyl)nitrobenzene, 2-(o-Nitrophenyl)ethyl bromide, o-Nitrophenethyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 108.00

CHF 108.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CHF 108.00

About This Item

Empirical Formula (Hill Notation):
C8H8BrNO2
CAS Number:
Molecular Weight:
230.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 108.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

bp

127-130 °C/1 mmHg

mp

34-38 °C

functional group

bromo
nitro

SMILES string

[O-][N+](=O)c1ccccc1CCBr

InChI

1S/C8H8BrNO2/c9-6-5-7-3-1-2-4-8(7)10(11)12/h1-4H,5-6H2

InChI key

SJLIPOVZERLCNL-UHFFFAOYSA-N

Application

Reactant for:
  • Reduction at carbon cathodes
  • N-(ureidoalkyl)benzylpiperidines as CCR3 receptors antagonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service