Skip to Content
Merck
All Photos(1)

Documents

688215

Sigma-Aldrich

[6,6]-Thienyl C61 butyric acid methyl ester

≥99%

Synonym(s):

[60]ThPCBM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H12O2S
Molecular Weight:
916.91
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥99%

form

powder

solubility

organic solvents: soluble

SMILES string

COC(=O)CCCC2(c1cccs1)[C]3=4c5c6c7c8c9c%10c(c%11c%12c3c%13c5c%14c%15c6c%16c7c%17c9c%18c%19c%10c%20c%11c%21c%12c%22c%13c%23c%14c%24c%15c%25c%16c%26c%17c%18c%27c%28c%19c%20c%29c%21c%30c%22c%23c%31c%24c%32c%25c%26c%27c%33c%28c%29c%30c%31c%32%33)[C]2=48

Application

[6,6]-Thienyl C61 butyric acid methyl ester is a class of thienyl based PCBM that can be used for the fabrication of optoelectronic devices such as organic solar cells (OSCs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Popescu, M.; van't Hof, P.; Sieval, A.; Jonkman, H.; Hummelen, J
Applied Physics Letters, 89, 213507-213507 (2006)
Electron trapping in higher adduct fullerene-based solar cells
Lenes M, et al.
Advances in Functional Materials, 19(18), 3002-3007 (2009)
Nanoscale electro-optical properties of organic semiconducting thin films: From individual materials to the blend
Escasain E, et al.
The Journal of Physical Chemistry C, 116(33), 17919-17927 (2012)

Articles

Progress in bulk heterojunction photodiodes since 1995 has improved device performance and widened research scope.

PCBM-based n-type semiconductors - Find p- and n-type organic semiconductors available with PCBM library & properties.

Find various photovoltaic and bioscience-based applications of fullerenes.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service