Skip to Content
Merck
All Photos(1)

Key Documents

683027

Sigma-Aldrich

trans-4-Fluorocinnamaldehyde

95%

Synonym(s):

(E)-3-(4-Fluorophenyl)-2-propenal

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 135.00

CHF 135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
CHF 135.00

About This Item

Empirical Formula (Hill Notation):
C9H7FO
CAS Number:
Molecular Weight:
150.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 135.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.599

density

1.146 g/mL at 25 °C

functional group

aldehyde
fluoro

SMILES string

Fc1ccc(\C=C\C=O)cc1

InChI

1S/C9H7FO/c10-9-5-3-8(4-6-9)2-1-7-11/h1-7H/b2-1+

InChI key

YSIYEWBILJZDQH-OWOJBTEDSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service