Skip to Content
Merck
All Photos(1)

Key Documents

682950

Sigma-Aldrich

1,1′-Bis[(11bR)-dinaphtho[2,1-d:1′, 2′-f][1,3,2]dioxaphosphepin-4-yl]ferrocene

Synonym(s):

R, R-Reetz F-Diphosphonite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H32FeO4P2
CAS Number:
Molecular Weight:
814.58
MDL number:
UNSPSC Code:
12352000

SMILES string

[Fe].[CH]1[CH][CH][C]([CH]1)P2Oc3ccc4ccccc4c3-c5c(O2)ccc6ccccc56.[CH]7[CH][CH][C]([CH]7)P8Oc9ccc%10ccccc%10c9-c%11c(O8)ccc%12ccccc%11%12

Application

Used in a variety of asymmetric reactions, including: hydrogenations and conjugate additions.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reetz, M. T.
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 71, 1503-1503 (1999)
Reetz, M. T. et al.
Chemical Communications (Cambridge, England), 2077-2077 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service