Skip to Content
Merck
All Photos(1)

Key Documents

681318

Sigma-Aldrich

N-Boc-piperidine-3-methanol

97%

Synonym(s):

1,1-Dimethylethyl 3-(hydroxymethyl)-1-piperidinecarboxylate, 3-(Hydroxymethyl)-N-(tert-butoxycarbonyl)piperidine, N-Boc-3-(hydroxymethyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO3
CAS Number:
Molecular Weight:
215.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

77-81 °C

SMILES string

CC(C)(C)OC(=O)N1CCCC(CO)C1

InChI

1S/C11H21NO3/c1-11(2,3)15-10(14)12-6-4-5-9(7-12)8-13/h9,13H,4-8H2,1-3H3

InChI key

OJCLHERKFHHUTB-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Pim-1 inhibitors
  • Vasopressin1b receptor antagonists
  • CXCR4 antagonists as anti-HIV agents
  • Amide CCR5 antagonist
  • PSSRI-based inhibitors of S. aureus multidrug efflux pumps
  • Human GnRH receptor antagonists

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service