Skip to Content
Merck
All Photos(3)

Key Documents

681067

Sigma-Aldrich

2-Aminothiophene-3-carbonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 51.80

CHF 51.80


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
CHF 51.80

About This Item

Empirical Formula (Hill Notation):
C5H4N2S
CAS Number:
Molecular Weight:
124.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 51.80


Estimated to ship on16 April 2025


Request a Bulk Order

Assay

97%

form

solid

mp

104-108 °C

functional group

nitrile

SMILES string

Nc1sccc1C#N

InChI

1S/C5H4N2S/c6-3-4-1-2-8-5(4)7/h1-2H,7H2

InChI key

XVGHZFWFGXDIOU-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service