Skip to Content
Merck
All Photos(1)

Documents

680001

Sigma-Aldrich

4-Methyl-3-thiopheneboronic acid pinacol ester

96%

Synonym(s):

4,4,5,5-Tetramethyl-2-(4-methyl-3-thienyl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(4-methylthien-3-yl)-1,3,2-dioxaborolane, 4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17BO2S
CAS Number:
Molecular Weight:
224.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.5039

bp

72-75 °C/0.2-0.3 mmHg

density

1.031 g/mL at 25 °C

SMILES string

Cc1cscc1B2OC(C)(C)C(C)(C)O2

InChI

1S/C11H17BO2S/c1-8-6-15-7-9(8)12-13-10(2,3)11(4,5)14-12/h6-7H,1-5H3

InChI key

XXSMGZQARORQMO-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service