Skip to Content
Merck
All Photos(2)

Key Documents

675180

Sigma-Aldrich

Isoquinoline-5-carboxaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO
CAS Number:
Molecular Weight:
157.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

116-120 °C

functional group

aldehyde

SMILES string

[H]C(=O)c1cccc2cnccc12

InChI

1S/C10H7NO/c12-7-9-3-1-2-8-6-11-5-4-10(8)9/h1-7H

InChI key

ILRSABOCKMOFGW-UHFFFAOYSA-N

Application

Direct conversion of heteroaryl-carboxaldehydes to acetonitriles by treatment with cyanoethylphosphonate followed by samarium(II) iodide reduction.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Neil Dalphin et al.
eNeuro, 7(3) (2020-04-24)
Membrane voltage oscillations in layer 1 (L1) of primary sensory cortices might be important indicators of cortical gain control, attentional focusing, and signal integration. However, electric field recordings are hampered by the low seal resistance of electrodes close to the
Tetrahedron Letters, 44, 2903-2903 (2003)

Articles

Synthesis of selective aldosterone synthase inhibitors using Wittig reaction with heterocyclic aldehydes described in study by Hartmann et al.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service