Skip to Content
Merck
All Photos(1)

Key Documents

673102

Sigma-Aldrich

Z-D-Prolinol

97%

Synonym(s):

(R)-2-Hydroxymethylpyrrolidine-1-carboxylic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO3
CAS Number:
Molecular Weight:
235.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.5396

density

1.135 g/mL at 25 °C

application(s)

peptide synthesis

SMILES string

OC[C@H]1CCCN1C(=O)OCc2ccccc2

InChI

1S/C13H17NO3/c15-9-12-7-4-8-14(12)13(16)17-10-11-5-2-1-3-6-11/h1-3,5-6,12,15H,4,7-10H2/t12-/m1/s1

InChI key

BJTNHGVCFWDNDP-GFCCVEGCSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service