Skip to Content
Merck
All Photos(1)

Key Documents

672084

Sigma-Aldrich

(S)-2-(Aminomethyl)-1-Boc-pyrrolidine

97%

Synonym(s):

(S)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, tert-Butyl (S)-2-(aminomethyl)-1-pyrrolidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
Beilstein:
8905233
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (GC)
96.5-103.5% (NT)
97%

form

solid

optical purity

enantiomeric excess: ≥97.5% (GC)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCC[C@H]1CN

InChI

1S/C10H20N2O2/c1-10(2,3)14-9(13)12-6-4-5-8(12)7-11/h8H,4-7,11H2,1-3H3/t8-/m0/s1

InChI key

SOGXYCNKQQJEED-QMMMGPOBSA-N

Application

(S)-2-(Aminomethyl)-1-Boc-pyrrolidine can be used as a building block to synthesize:
  • Imidazo[1,2-b]pyridazine derivatives as potent IKKβ inhibitors.[1]
  • 1,2-dicarba-closo-dodecaborane (o-carborane) and 1,7-dicarba-closo-dodecaborane (m-carborane) derivatives as potential D2 receptor antagonists.[2]
  • Pyrrolidine based Merrifield resin as a chiral organocatalyst for the asymmetric Michael addition reaction of ketones with nitrostyrenes.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of D2 receptor ligand analogs incorporating one dicarba-closo-dodecaborane unit
Vazquez, N, et al.
Tetrahedron Letters, 52(5), 615-618 (2011)
Discovery of imidazo [1, 2-b] pyridazines as IKK? inhibitors. Part 3: Exploration of effective compounds in arthritis models
Shimizu H, et al.
Bioorganic & Medicinal Chemistry Letters, 21(15), 4550-4555 (2011)
Recyclable Merrifield resin-supported organocatalysts containing pyrrolidine unit through A3-coupling reaction linkage for asymmetric Michael addition
Liu J, et al.
Chirality, 22(4), 432-441 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service