Skip to Content
Merck
All Photos(1)

Key Documents

671649

Sigma-Aldrich

(1S,2S)-1-Amino-2-benzyloxycyclopentane

98%

Synonym(s):

(1S)-trans-2-(Phenylmethoxy)cyclopentanamine, (1S,2S)-trans-2-Benzyloxycyclopentylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
Beilstein:
9554058
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.5% (GC)
98%

optical purity

enantiomeric excess: ≥98%

impurities

≤5% water

storage temp.

2-8°C

SMILES string

N[C@H]1CCC[C@@H]1OCc2ccccc2

InChI

1S/C12H17NO/c13-11-7-4-8-12(11)14-9-10-5-2-1-3-6-10/h1-3,5-6,11-12H,4,7-9,13H2/t11-,12-/m0/s1

InChI key

JIMSXLUBRRQALI-RYUDHWBXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(1S,2S)-1-Amino-2-benzyloxycyclopentane can be used:
  • As a component of the ligand cocktail, which is used in the study of binding and inhibitory activities of PA4794 protein.[1]
  • In one of the key synthetic steps for the preparation of benzoxazole based mPGES-1 inhibitors[2] and trans-2-aminocyclopentanol derivatives.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural, functional and inhibition studies of a GNAT superfamily protein PA4794: a new C-terminal lysine protein acetyltransferase from Pseudomonas aeruginosa
Majorek KA, et al.
Test, 30223-30235 (2013)
Novel benzoxazole inhibitors of mPGES-1
Kablaoui N, et al.
Bioorganic & Medicinal Chemistry Letters, 23(3), 907-911 (2013)
Structural, functional and inhibition studies of a GNAT superfamily protein PA4794: a new C-terminal lysine protein acetyltransferase from Pseudomonas aeruginosa
Majorek KA, et al.
The Journal of Biological Chemistry, 30223-30235 (2013)
Structural, functional and inhibition studies of a GNAT superfamily protein PA4794: a new C-terminal lysine protein acetyltransferase from Pseudomonas aeruginosa
Majorek KA, et al.
The Journal of biological chemistry, 30223-30235 (2013)
Highly efficient chemoenzymatic syntheses of trans-2-aminocyclopentanol derivatives
Gonzalez-Sabin J, et al.
Journal of Molecular Catalysis. B, Enzymatic, 59(1-3), 111-115 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service