Skip to Content
Merck
All Photos(1)

Documents

667226

Sigma-Aldrich

Chlorohydridotris(triphenylphosphine) ruthenium(II) toluene adduct

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HRuCl · 3(P(C6H5)3) ·C6H5CH3
CAS Number:
Molecular Weight:
1016.53
UNSPSC Code:
12161600
PubChem Substance ID:

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

storage temp.

2-8°C

SMILES string

Cl[RuH].Cc1ccccc1.c2ccc(cc2)P(c3ccccc3)c4ccccc4.c5ccc(cc5)P(c6ccccc6)c7ccccc7.c8ccc(cc8)P(c9ccccc9)c%10ccccc%10

InChI

1S/3C18H15P.C7H8.ClH.Ru.H/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;;;/h3*1-15H;2-6H,1H3;1H;;/q;;;;;+1;/p-1

InChI key

FPDVFAUPHLXLMN-UHFFFAOYSA-M

Application

Reactant for synthesis of:
  • Dithiolate-bridged ruthenium-rhodium heterobimetallic dihydride complex
  • Amido / phosphine pincer hydrides of ruthenium

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service