Skip to Content
Merck
All Photos(1)

Key Documents

662461

Sigma-Aldrich

Lumidot CdSe

640, core-type quantum dots, 5 mg/mL in toluene

Synonym(s):

Cadmium monoselenide, Cadmium selenide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdSe
CAS Number:
Molecular Weight:
191.37
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Quality Level

form

dispersion

concentration

5 mg/mL in toluene

particle size

6.2-7.7 nm

λmax

615-625 nm

fluorescence

λem 635-645 nm

storage temp.

2-8°C

InChI

1S/Cd.Se

InChI key

AQCDIIAORKRFCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Organically stabilized cadmium selenide quantum dots in toluene

Analysis Note

Particle size was determined by TEM according to the procedure of Yu, W. W., et al., Chem. Mater., 2003, 15, 2854.
Quantum yield was measured following the procedure of Qu, L., Peng, X., J. Am. Chem. Soc., 2002, 124-2049.

Other Notes

Lifetime one year if stored properly

Legal Information

Product of Nanoco Technologies sold under US and worldwide patents and patents pending.
Lumidot is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan T Spann et al.
Optics express, 21 Suppl 1, A15-A22 (2013-02-15)
Ultrafast time-resolved absorption spectroscopy is used to investigate exciton dynamics in CdSe nanocrystal films. The effects of morphology, quantum-dot versus quantum-rod, and preparation of nanocrystals in a thin film form are investigated. The measurements revealed longer intraband exciton relaxation in
Svetlana V Kilina et al.
Physical review letters, 110(18), 180404-180404 (2013-05-21)
Quantum confinement can dramatically slow down electron-phonon relaxation in nanoclusters. Known as the phonon bottleneck, the effect remains elusive. Using a state-of-the-art time-domain ab initio approach, we model the observed bottleneck in CdSe quantum dots and show that it occurs
Shancheng Yan et al.
Journal of nanoscience and nanotechnology, 13(1), 23-32 (2013-05-08)
Semiconducting nanowire heterostructures are of particular interest because of their fascinating properties and potential applications in the field of nanoscale science. CdS, with a direct bandgap of 2.42 eV, is considered to be an excellent material for various optoelectronic applications
Yuanyuan Wang et al.
Inorganic chemistry, 52(6), 2933-2938 (2013-03-06)
Four [(CdSe)13(RNH2)13] derivatives (R = n-propyl, n-pentyl, n-octyl, and oleyl) are prepared by reaction of Cd(OAc)2·2H2O and selenourea in the corresponding primary-amine solvent. Nanoclusters grow in spontaneously formed amine-bilayer templates and are characterized by elemental analysis, IR spectroscopy, UV-vis spectroscopy
David T R Stewart et al.
Environmental toxicology and chemistry, 32(6), 1288-1294 (2013-02-19)
The present study addresses the bioaccumulation behavior of cadmium selenide quantum dots by Eisenia andrei earthworms in a terrestrial environment. Earthworms were exposed to quantum dot-treated soil for up to 4 wk and analyzed for cadmium and selenium concentration using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service