Skip to Content
Merck
All Photos(1)

Key Documents

656720

Sigma-Aldrich

2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3Cl2N3O
CAS Number:
Molecular Weight:
192.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

208-224 °C

functional group

aldehyde
chloro

storage temp.

2-8°C

SMILES string

[H]C(=O)c1c(Cl)nc(N)nc1Cl

InChI

1S/C5H3Cl2N3O/c6-3-2(1-11)4(7)10-5(8)9-3/h1H,(H2,8,9,10)

InChI key

GOJUJUVQIVIZAV-UHFFFAOYSA-N

Application

Substrate used in the synthesis of an N-terminal surrogate in amino acid and peptide analogues.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 3159-3159 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service