Skip to Content
Merck
All Photos(1)

Documents

65569

Sigma-Aldrich

Methylamine

≥99.0%

Synonym(s):

Monomethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH2
CAS Number:
Molecular Weight:
31.06
Beilstein:
741851
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

1.08 (20 °C, vs air)

vapor pressure

27 psi ( 20 °C)

Assay

≥99.0%

autoignition temp.

806 °F

expl. lim.

4.9-20.8 %

impurities

≤0.05% ammonia
≤0.05% trimethylamine
≤0.2% dimethylamine

bp

-6.3 °C (lit.)

mp

-93 °C (lit.)

density

0.70 g/mL at 20 °C (lit.)

SMILES string

CN

InChI

1S/CH5N/c1-2/h2H2,1H3

InChI key

BAVYZALUXZFZLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin with 100 mL (net ~70 g)

Other Notes

Sales restrictions may apply

control valve

Product No.
Description
Pricing

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Compr. Gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Yu Chin et al.
Nature communications, 6, 7383-7383 (2015-06-26)
Despite the widespread use of solution-processable hybrid organic-inorganic perovskites in photovoltaic and light-emitting applications, determination of their intrinsic charge transport parameters has been elusive due to the variability of film preparation and history-dependent device performance. Here we show that screening
Jeff O'Sullivan et al.
Neurotoxicology, 25(1-2), 303-315 (2003-12-31)
The semicarbazide-sensitive amine oxidases (SSAO) (EC 1.4.3.6) were believed to be detoxifying enzymes, primarily involved in the oxidative deamination of endogenous amines, such as methylamine and aminoacetone, together with some xenobiotic amines. However, it appears that the reaction products may
T Kokubo et al.
Journal of biomedical materials research, 24(6), 721-734 (1990-06-01)
High-strength bioactive glass-ceramic A-W was soaked in various acellular aqueous solutions different in ion concentrations and pH. After soaking for 7 and 30 days, surface structural changes of the glass-ceramic were investigated by means of Fourier transform infrared reflection spectroscopy
Katja Wosikowski et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 9(5), 1917-1926 (2003-05-10)
To avoid systemic toxicity of the cytotoxic drug methotrexate (MTX) and to improve tumor selectivity, MTX was bound to human serum albumin (HSA) as a drug carrier. To understand more about the mechanism of action of MTX conjugated to HSA
Liangcai Zhao et al.
Journal of ethnopharmacology, 142(3), 647-656 (2012-06-13)
Zhibai Dihuang Pill (ZDP) is one of ancient traditional Chinese medicines (TCMs), which is usually used for the treatment of kidney deficiency for thousands of years in China. Traditional Chinese medicines (TCMs) usually operate in vivo through multi-components, multi-ways and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service