Skip to Content
Merck
All Photos(3)

Key Documents

647403

Sigma-Aldrich

2-Chloro-4-iodopyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3ClIN
CAS Number:
Molecular Weight:
239.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

42-43 °C (lit.)

functional group

chloro
iodo

SMILES string

Clc1cc(I)ccn1

InChI

1S/C5H3ClIN/c6-5-3-4(7)1-2-8-5/h1-3H

InChI key

KJKIPRQNFDUULB-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyomin Jin et al.
Molecules (Basel, Switzerland), 24(1) (2019-01-10)
Herein, we investigated the effect of ring planarity by fully characterizing four pyridine-based o-carboranyl compounds. o-Carborane was introduced to the C4 position of the pyridine rings of 2-phenylpyridine and 2-(benzo[b]thiophen-2-yl)pyridine (CB1 and CB2, respectively), and the compounds were subsequently borylated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service