Skip to Content
Merck
All Photos(1)

Key Documents

637815

Sigma-Aldrich

2-Bromoiodobenzene-13C6

≥99 atom % 13C, ≥99% (CP), contains copper as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H4BrI
CAS Number:
Molecular Weight:
288.86
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

contains

copper as stabilizer

bp

120-121 °C/15 mmHg (lit.)

mass shift

M+6

SMILES string

Br[13c]1[13cH][13cH][13cH][13cH][13c]1I

InChI

1S/C6H4BrI/c7-5-3-1-2-4-6(5)8/h1-4H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

OIRHKGBNGGSCGS-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service