Skip to Content
Merck
All Photos(1)

Key Documents

616273

Sigma-Aldrich

DL-Phenyl-d5-alanine

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 742.00

CHF 742.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
CHF 742.00

About This Item

Linear Formula:
C6D5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
170.22
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

CHF 742.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

solid

mp

266-267 °C (dec.) (lit.)

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(CC(N)C(O)=O)c([2H])c1[2H]

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/i1D,2D,3D,4D,5D

InChI key

COLNVLDHVKWLRT-RALIUCGRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zhang et al.
Electrophoresis, 40(18-19), 2309-2320 (2019-04-27)
The actual utility of capillary electrophoresis-mass spectrometry (CE-MS) for biomarker discovery using metabolomics still needs to be assessed. Therefore, a simulated comparative metabolic profiling study for biomarker discovery by CE-MS was performed, using pooled human plasma samples with spiked biomarkers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service