Skip to Content
Merck
All Photos(1)

Key Documents

612502

Supelco

1,2-Dichlorobenzene solution

NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
CHF 2’370.00

CHF 2’370.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 EACH
CHF 2’370.00

About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

CHF 2’370.00


Check Cart for Availability

Request a Bulk Order

grade

NMR reference standard
analytical standard

concentration

1% in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

SMILES string

Clc1ccccc1Cl

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

RFFLAFLAYFXFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

1H resolution

Quantity

3 mm O.D. tube contains 0.235 mL.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service