604089
Chloroacetyl chloride-13C2
99 atom % 13C, 99% (CP)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Cl13CH213COCl
CAS Number:
Molecular Weight:
114.93
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
Assay
99% (CP)
bp
105-106 °C (lit.)
mp
-22 °C (lit.)
density
1.443 g/mL at 25 °C
mass shift
M+2
SMILES string
Cl[13CH2][13C](Cl)=O
InChI
1S/C2H2Cl2O/c3-1-2(4)5/h1H2/i1+1,2+1
InChI key
VGCXGMAHQTYDJK-ZDOIIHCHSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT RE 1
Target Organs
Lungs
Supplementary Hazards
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
212.0 °F - closed cup
Flash Point(C)
100 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service