Skip to Content
Merck
All Photos(2)

Key Documents

590770

Sigma-Aldrich

2-(2-Thienyl)furan

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6OS
CAS Number:
Molecular Weight:
150.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.6210 (lit.)

bp

209 °C (lit.)

density

1.192 g/mL at 25 °C (lit.)

SMILES string

c1coc(c1)-c2cccs2

InChI

1S/C8H6OS/c1-3-7(9-5-1)8-4-2-6-10-8/h1-6H

InChI key

RVYKTPRETIBSPK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

198.0 °F

Flash Point(C)

92.2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Wen Kuo et al.
Membranes, 11(2) (2021-02-13)
Five carbazole-containing polymeric membranes (PDTC, P(DTC-co-BTP), P(DTC-co-BTP2), P(DTC-co-TF), and P(DTC-co-TF2)) were electrodeposited on transparent conductive electrodes. P(DTC-co-BTP2) shows a high ΔT (68.4%) at 855 nm. The multichromic properties of P(DTC-co-TF2) membrane range between dark yellow, yellowish-green, gunmetal gray, and dark

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service