Skip to Content
Merck
All Photos(1)

Key Documents

586897

Sigma-Aldrich

Allyl 2-bromobenzyl ether

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2OCH2CH=CH2
CAS Number:
Molecular Weight:
227.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.5460 (lit.)

bp

251-252 °C (lit.)

density

1.327 g/mL at 25 °C (lit.)

SMILES string

Brc1ccccc1COCC=C

InChI

1S/C10H11BrO/c1-2-7-12-8-9-5-3-4-6-10(9)11/h2-6H,1,7-8H2

InChI key

WBVJSBVYPDCMEW-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher L Rock et al.
Dalton transactions (Cambridge, England : 2003), 48(2), 461-467 (2018-11-30)
The phosphine-substituted α-diimine Ni precursor, (Ph2PPrDI)Ni, has been found to catalyze alkene hydrosilylation in the presence of Ph2SiH2 with turnover frequencies of up to 124 h-1 at 25 °C (990 h-1 at 60 °C). Moreover, the selective hydrosilylation of allylic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service