Skip to Content
Merck
All Photos(1)

Key Documents

579351

Sigma-Aldrich

Poly(2-vinylnaphthalene)

average Mn ~20,000

Synonym(s):

2-Vinylnaphthalene polymer, Poly(β-vinylnaphthalene), Poly(2-vinylnaphthalene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C10H7)]n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

mol wt

Mw/Mn 1.15
average Mn ~20,000

transition temp

Tg (DSC) 141 °C (onset)

InChI

1S/C12H10/c1-2-10-7-8-11-5-3-4-6-12(11)9-10/h2-9H,1H2

InChI key

KXYAVSFOJVUIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Narrow molecular weight distribution.

Preparation Note

Prepared by anionic polymerization.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengyang Wang et al.
Macromolecular rapid communications, 33(10), 933-937 (2012-03-01)
A straightforward strategy for assembling polymeric dimers from amphiphilic nanoparticles is reported. Amphiphilic polymeric nanoparticles with a mixed-shell of PEO/P2VN blocks and a flexible core of PAA blocks are fabricated by a non-covalent crosslinking method. Uniform polymeric dimers are efficiently
W J Litchfield et al.
Clinical chemistry, 30(9), 1489-1493 (1984-09-01)
Recent innovations in particle design have led to the development of highly sensitive and reproducible immunoassay methods for the Du Pont aca discrete clinical analyzer. Key advances include the synthesis and use of particles less than 1 micron in diameter

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service