Skip to Content
Merck
All Photos(1)

Key Documents

574945

Sigma-Aldrich

Hexaamminenickel(II) bromide

99.999%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ni(NH3)6]Br2
CAS Number:
Molecular Weight:
320.68
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.999%

form

solid

reaction suitability

reagent type: catalyst
core: nickel

SMILES string

N.N.N.N.N.N.Br[Ni]Br

InChI

1S/2BrH.6H3N.Ni/h2*1H;6*1H3;/q;;;;;;;;+2/p-2

InChI key

GDRWEJQXKZQPJA-UHFFFAOYSA-L

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service