All Photos(1)
About This Item
Linear Formula:
FC6H3(OH)CO2H
CAS Number:
Molecular Weight:
156.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
214-218 °C (lit.)
SMILES string
OC(=O)c1ccc(F)c(O)c1
InChI
1S/C7H5FO3/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,9H,(H,10,11)
InChI key
QATKOZUHTGAWMG-UHFFFAOYSA-N
Application
4-Fluoro-3-hydroxybenzoic acid may serve as a starting material in the synthesis of the following potent inhibitors of β-arylsulfotransferase IV (β-AST-IV):
- 6-(2-(N-4-fluoro-3-hydroxybenzoylamino)ethylamino)purine via reaction with O-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate(HBTU) and 6-((2-ammonium trifluroacetate)ethylamino)purine
- 6-(12-(N-4-fluoro-3-hydroxybenzoylamino)dodecylamino)purine via reaction with O-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate(HBTU) and 6-((12-ammonium trifluroacetate)dodecylamino)purine
- 6-(12-(N-4-fluoro-3-hydroxybenzoylamino)dodecylamino)purine via reaction with O-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate(HBTU) and 6-((12-ammonium trifluroacetate)dodecylamino)purine
- 2-chloro-6-(12-(N-4-fluoro-3-hydroxybenzoylamino)dodecylamino)purine via reaction with O-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate(HBTU) and 2-chloro-6-((12-ammonium trifluroacetate)dodecylamino)purine
- 12-N-boc-1-N-(4-fluoro-3-hydroxybenzoyl)diaminododecane via reaction with O-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate(HBTU) and N-boc-1,12-diaminododecane
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
"Rapid Discovery of Potent Sulfotransferase Inhibitors by Diversity-Oriented Reaction in Microplates Followed by in situ Screening"
Dr D,BM, et al.
Chembiochem, 5(6), 811-819 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service