Skip to Content
Merck
All Photos(1)

Key Documents

568325

Sigma-Aldrich

Ethyl 3-chloro-4-hydroxybenzoate

97%

Synonym(s):

3-Chloro-4-hydroxybenzoicacid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3ClCO2CH2CH3
CAS Number:
Molecular Weight:
200.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

98-102 °C (lit.)

SMILES string

CCOC(=O)c1ccc(O)c(Cl)c1

InChI

1S/C9H9ClO3/c1-2-13-9(12)6-3-4-8(11)7(10)5-6/h3-5,11H,2H2,1H3

InChI key

QBOWIPYEPOVPGR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhui Li et al.
Environmental science and pollution research international, 22(22), 17987-17997 (2015-07-15)
As an emerging group of endocrine-disrupting chemicals, parabens have attracted growing attention due to their potential effects on human health. In the present study, the occurrence and distribution of eight parabens, four chlorinated parabens, and their common hydrolysis product, p-hydroxybenzoic
Wenhui Li et al.
The Science of the total environment, 539, 262-270 (2015-09-13)
The occurrence and distribution of 13 target compounds, including eight parabens, four chlorinated parabens and p-hydroxybenzoic acid (PHBA), were detected in surface water samples at 35 sampling sites in the Beijing River system, China. The surface water samples were collected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service